Learn More
Market Lab Marketlab Mobile Draw Station Plus with Tray
Supplier:  Market Lab 59758
Encompass_Preferred
The Marketlab Mobile Draw Station Plus is a mobile phlebotomy cart designed for blood collection and processing. It features a stainless steel work surface, storage for supplies, a sharps container, a waste receptacle, and a blood tube rocker. The cart has 5" casters for easy mobility and can be customized with different drawer configurations. This phlebotomy cart provides efficient workflow and organization for blood draws and processing blood samples.
Catalog No. NC1975635
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.