Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Mobile Tables

Catalog No. 05413976 Shop All Eppendorf North America Products
Change view
Click to view available options
Product Type:
High Version
Low Version
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
05-413-976 High Version
05-413-975 Low Version
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-413-976 Supplier Eppendorf™ Supplier No. 5921000012
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eppendorf™ Mobile Tables ideal for ergonomic replacement of floor-standing centrifuges

  • Fits in any empty floor space
  • Available in two different height options
  • Ideal for replacing a floor-standing centrifuge
  • Ready to use - no assembly necessary
  • Dimensions (W x D x H): approx. 75 x 70 x 58.5 cm
  • For use with Centrifuges 5804/5804 R, 5810/5810 R, 5910 R and 5920 R
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Centrifuge 5804/5804 R, 5810/5810 R, 5910 R, 5920 R
Length (Metric) 70 cm
Height (Metric) 58.5 cm
Width (Metric) 75 cm
Includes Heavy-Duty Transport Wheels and Brakes
Product Type High Version
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.