Learn More
Apexbio Technology LLC Moclobemide (Ro 111163),10mg CAS# 71320-77-9
SDP
Supplier:  Apexbio Technology LLC A437010
Encompass_Preferred
Moclobemide is a reversible inhibitor of monoamine oxidase-A (MAO-A) with IC50 value of 10μM [1].Moclobemide is a prototype of RIMA agents. It is found to possess antidepressant efficacy with less risk of fatal side-effects like hypertensive crisis. Moclobemide shows a weak but specific inhibition of MAO-A in the in vitro assay using rat brain homogenates. The IC50 values of MAO-A and MAO-B in the assay are 6mM and 1000mM, respectively. In ex vivo animal experiments, moclobemide produces an inhibition of 80% in the brain and liver. Additionally, the long-term administration of high doses of moclobemide is found to down-regulate β-adrenoceptors and increase the agonist binding affinity of α1-adrenoceptors. Moreover, moclobemide is rapidly and almost completely absorbed after oral administration. The bioavailability of moclobemide is about 50% after a single administration of 100 mg [2]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0556
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.