Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Model Human Skull - Numbered 3 Parts PROMO
SCI_ED

Catalog No. S15581PM Shop All Eisco Products
Change view
Click to view available options
Dimensions (L x W x H):
8.0 x 6.0 x 8.0 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S15581PM 8.0 x 6.0 x 8.0 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15581PM Supplier Eisco™ Supplier No. AM0124ASPROMO/DEL

Human skull model replica.

  • Each part on the skull is numbered
  • Jaw is spring loaded
  • Removable Skull cap
  • Skull is made of durable plastic

Specifications

Dimensions (L x W x H) 8.0 x 6.0 x 8.0 in.
Includes Laminated sheet with each numbers name for identification
Material Durable Plastic
Weight (English) 2.2 lb.
Product Type Human Skull Model
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.