Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Model Plus The Flower
SCI_ED

Catalog No. S94388 Shop All Si Metric Manufacturing Limited Products
Change view
Click to view available options
Grade:
1 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S94388 1 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Catalog No. S94388 Supplier Si Metric Manufacturing Limited Supplier No. 69070

missing translation for 'validMainframeMsg'

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Includes

  • Detailed teacher's guide with complete background information, reproducible worksheets, cross-curricular extension activities and assessment ideas

Colorful teaching model.

Designed specifically for hands-on use with students of all ages.

  • Made of resilient, nontoxic EVA foam
  • Removable pieces stimulate problem-solving skills and develop small-motor skills
  • Dimensions: 19.5 x 19.5 x 15 in. (49.5 x 49.5 x 38.1cm)

Specifications

Grade 1 to 8
Format Model
Product Type Model Plus Flowers
Includes Teachers guide with background information, reproducible worksheets, extension activities
For Use With (Application) Designed specifically for hands-on use with students of all ages
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.