Learn More
Description
- Permanganate step is omitted, only two step staining technique for acid fast organisms saves lot of time
- For all non-acid fast organisms and debris, new counterstain quenches fluorescence
- Yields cleaner field of view
Specifications
Specifications
| Quantity | 1 x 4L |
| Includes | Stain and decolorizer or quencher |
| Format | Bottle |
| For Use With (Application) | Rapid and economical staining technique for acid fast organisms |
| Product Type | Modified Auramine O Stain |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
