Learn More
StatLab MODIFIED BIELSCHOWSKY'S KIT
SDP
Supplier:  StatLab KTBIE
Encompass_Preferred
This kit stains for axons, intracellular neurofibrils, and senile plaques in 35 minutes. Stain kit includes Silver Nitrate Solution, 5% Sodium Thiosulfate, Stock Developer Solution A, B, and C.
Catalog No. NC9765117
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.