Learn More
Epredia™ Modified Grocott' s Methenamine Silver (Periodic Acid) Stain Kit
Catalog No. 88024 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Fast Green Stain Solution
Gold Chloride Solution (0.1%)
Silver Stain Kit, Periodic Acid
Sodium Thiosulfate Solution
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
88-024 Fast Green Stain Solution
23-900-656 Silver Stain Kit, Periodic Acid
88-025 Sodium Thiosulfate Solution
22-110-652 Gold Chloride Solution (0.1%)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 88-024 Supplier Epredia™ Supplier No. 88024
Add to Cart
Add to Cart
This item is not returnable. View return policy
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid)
  • Fungi, basement membranes and opportunistic organisms, along with reticular fibers, are stained black
  • Background is stained green
Specifications
Quantity 125 mL
Product Type Fast Green Stain Solution