Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Modular Blocks and Accessories
SCI_ED

Catalog No. S38776 Shop All OHAUS Products
Change view
Click to view available options
Sample Type:
1.5mL Tube
10mm Tube
15/16mm Tube
15mL Tube
50mL Tube
Type:
Dry Block
Temperature Probe
Depth (Metric) Well:
3.91 cm
4.45 cm
4.76 cm
4.8 cm
Diameter (Metric) Well:
1.07 cm
1.1 cm
1.71 cm
1.75 cm
2.9 cm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type Type Depth (Metric) Well Diameter (Metric) Well
S38776 50mL Tube Dry Block 4.76 cm 2.9 cm
S38779 1.5mL Tube Dry Block 3.91 cm 1.1 cm
S38777 15mL Tube Dry Block 4.45 cm 1.71 cm
S38774 15/16mm Tube Dry Block 4.8 cm 1.75 cm
S38775 10mm Tube Dry Block 4.8 cm 1.07 cm
S38772 - Temperature Probe - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
To avoid possible electrical hazard, DO NOT fill well or block with water or other liquids. Units are designed as a dry bath/incubator.
Catalog No. S38776 Supplier OHAUS™ Supplier No. 30400168
Add to Cart
Add to Cart
To avoid possible electrical hazard, DO NOT fill well or block with water or other liquids. Units are designed as a dry bath/incubator.

Constructed from solid anodized aluminum block.

Modular blocks are sold separately from heaters.

  • Close contact of tubes-to-block-walls allow for maximum heat retention
  • Each block has a thermometer well for measuring block temperature
TRUSTED_SUSTAINABILITY

Specifications

Description Module Block 50 mL Conical Centrifuge
For Use With (Equipment) Dry Block Heater
Material Anodized Aluminum
No. of Places 5
Type Dry Block
Sample Type 50mL Tube
Length (English) 3.75 in.
Length (Metric) 9.5 cm
Height (English) 2 in.
Height (Metric) 5.1 cm
Depth (English) Well 1.87 in.
Depth (Metric) Well 4.76 cm
Diameter (English) Well 1.14 in.
Diameter (Metric) Well 2.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.