Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Modular Blocks and Accessories

Catalog No. p-7184928 Shop All OHAUS Products
Change view
Click to view available options
Sample Type:
Titer Plates
Vials - 19 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
02-106-1016 Vials - 19 mm
02-106-1017 Titer Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator.
Catalog No. 02-106-1016 Supplier OHAUS™ Supplier No. 30400185
Add to Cart
Add to Cart
To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator.

OHAUS™ Modular Blocks and Accessories are available in two different varieties, both are constructed from solid anodized aluminum block

  • Modular blocks are sold separately from heaters
  • Close contact of tubes-to-block-walls allow for maximum heat retention
  • Each block has a thermometer well for measuring block temperature
  • Available in single or double block options
  • Recessed well for better stability, flat surface for flat and round bottom plates
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Dry Block Heater
Material Anodized Aluminum
No. of Places 12
Type Module Block
Sample Type Vials - 19 mm
Size 1 Block
Length (English) 3.75 in.
Length (Metric) 9.5 cm
Height (English) 2 in.
Height (Metric) 5.1 cm
Depth (English) Well 1.77 in.
Depth (Metric) Well 4.5 cm
Diameter (English) Well 0.77 in.
Diameter (Metric) Well 1.97 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.