Learn More
Poltex Modular Serological Pipette Holder Starter Set
SDP
Catalog No. 22000661
Change view
Click to view available options
Positions:
1
2
Includes:
Pipette Controller Bracket
Without Bracket
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions Includes
22-000-661 1 Without Bracket
22-000-662 1 Pipette Controller Bracket
22-000-663 2 Without Bracket
22-000-664 2 Pipette Controller Bracket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-000-661 Supplier Poltex Supplier No. SRPIPSTAK1START
Add to Cart
Add to Cart
This item is not returnable. View return policy
Modular Serological Pipette Holder Starter Set - Available as a starter pack as shown in the image, with or without pipette controller bracket.
  • Modular serological pipette storage system.
  • Combine 2.5, 3.5, and/or 4.5 in. tall units in any number or combination to achieve your perfect storage system.
  • Durable and cleanable PETG construction.
  • Sold as a starter pack as shown in the image, with or without pipette controller bracket.
  • Pipette Controller Bracket (BRK) can be added (mounts to either side in multiple possible positions).
  • Add additional 2.5, 3.5, or 4.5 in. tall units.
  • Combine as many units in any combination for the perfect setup.
  • Hardware kits included with each unit.
  • Additional units are purchased as a separate product (SRPIPSTAK125, SRPIPSTAK135, and SRPIPSTAK145).
  • Hardware straps either join one unit to the next, or the bottom unit serves as a foot to elevate the front and provide a backward tilt.
  • See image for dimensions.
Specifications
Color Clear
Material PETG
Positions 1
Stackable Yes
Includes Without Bracket
Product Type Pipet Rack