Learn More
Poltex Modular Serological Pipette Holder Starter Set
Description
- Modular serological pipette storage system.
- Combine 2.5, 3.5, and/or 4.5 in. tall units in any number or combination to achieve your perfect storage system.
- Durable and cleanable PETG construction.
- Sold as a starter pack as shown in the image, with or without pipette controller bracket.
- Pipette Controller Bracket (BRK) can be added (mounts to either side in multiple possible positions).
- Add additional 2.5, 3.5, or 4.5 in. tall units.
- Combine as many units in any combination for the perfect setup.
- Hardware kits included with each unit.
- Additional units are purchased as a separate product (SRPIPSTAK125, SRPIPSTAK135, and SRPIPSTAK145).
- Hardware straps either join one unit to the next, or the bottom unit serves as a foot to elevate the front and provide a backward tilt.
- See image for dimensions.
Specifications
Specifications
| Color | Clear |
| Material | PETG |
| Positions | 2 |
| Stackable | Yes |
| Includes | Without Bracket |
| Product Type | Pipet Rack |