Learn More
Cole-Parmer Fluke 80TK Thermocouple Module for 180-series multimeter
Supplier: Cole-Parmer 2605263
This thermocouple module converts a DMM to a thermometer. It can be used with type-K thermocouples in low voltage applications that are below 24 V AC, 60 V DC. It has a measurement range of -50 to 1,000 degrees Celsius (probe dependent), and one year warranty.