Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Molded Rubber Kneepads

Impacto™ 885-00 Heavy duty rubber Kneepads provide protection from abrasion and cumulative trauma

Supplier:  Impacto™ 88500000000

Catalog No. 19-400-105


Only null left
Add to Cart

Description

Description

Impacto™ Rubber Kneepads

  • Molded, seamless design cups the kneecap
  • Heavy duty, crush proof, durable
  • Easy on/off replaceable and adjustable rubber straps fasten to plastic buttons
  • Great for work in wet environments

Compliance

CE compliance

Specifications

Specifications

7 x 7 x 2 in.
Molded rubber kneepads
6 in.
Industrial and construction settings, wet environments
1
Adjustable strap attaches to plastic button
15.24 cm
Universal
Molded rubber
Rubber
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.