Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ 9000 Series Full Face Respirator Facepiece
SDP

Catalog No. 19384316 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Format:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
19-384-316 Medium
19-384-315 Small
19-384-317 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-384-316 Supplier Moldex™ Supplier No. 9002
Add to Cart
Add to Cart
This item is not returnable. View return policy

At less than 13 ounces, the 9000 Series respirator is ultra-lightweight. Workers will appreciate the comfort and longer wearability and employers will benefit from increased user acceptance.

  • A convenient ratchet fastening strap system and the stand-away head harness make putting the 9000 mask on and taking it off quick and easy. The strap tabs are securely molded directly to the flange for rugged use.
  • The 9000 boasts an extra-wide viewing lens, with an exclusive over-molded flange design. This eliminates the traditional clamping frame found on most other face pieces.
  • The 9000 is completely free of metal parts, making it ideal for mines, refineries and other work environments where metal may be hazardous.
  • Advanced design eliminates unnecessary parts. Moldex™'s overmolded flange and harness tabs, plus integrated exhalation assembly, make cleaning and maintaining the 9000 Series respirator a breeze.

Compliance

Lens meets ANSI™ Z87+.

Specifications

Certifications/Compliance ANSI Z87+
Material Polycarbonate Lens
Product Type Respirator Facepiece
Format Medium
For Use With (Application) Mines, refineries and other industries where metal parts can create an explosion hazard
Harness Style Stand away
Includes Tear-away lens protector
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.