Learn More
Moldex™ 9000 Series Respirator Nose Cup
Nose cup accessory for use with the 9000 Series full face respirator.
Supplier: Moldex™ 0094
This item is not returnable.
View return policy
Description
- Accessory for use with 9000 Series full face respirator
- Packed 5 per box
- Available in two sizes, small or medium/large
Specifications
Nosecup Assembly | |
Prevention of lens fogging and the removal of exhaled air | |
Two inhalation diaphragms |
Medium/Large | |
9000 series full face respirator | |
0094 |