Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Moldex™ Glide™ Foam No Roll Twist-In Ear Plugs

Glide™ Trio’s unique multi-curved stem rotates for a custom fit inside each ear canal. Simply push and twist to angle desired fit.
$155.70 - $172.40
Specifications
ANSI S3.1974 | Yes |
---|---|
CSA Z94.2 | Class AL |
Size | Universal |
For Use With (Application) | Construction, Utilities, Manufacturing, Production, Mining, Oil and Gas, and Other Various Industries |
High Visibility | No |
Catalog Number | Mfr. No. | Corded | Plug Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Corded | Plug Color | Price | Quantity & Availability | ||||
19-384-357
![]() |
Moldex™
6945 |
Corded | Green |
Pack of 100 for $172.40
Case of 4 PK for $655.00
|
|
||||
19-384-366
![]() |
Moldex™
6946 |
Uncorded | Camo |
Each for $155.70
Case of 4 EA for $592.00
|
|
||||
19-384-365
![]() |
Moldex™
6941 |
Corded | Camo | N/A | N/A | N/A | |||
19-384-356
![]() |
Moldex™
6940 |
Uncorded | Green | N/A | N/A | N/A | |||
Description
- Unique multi-curved stem rotates for a custom fit
- Large handle on curved stem for easy insertion and removal
- No roll down design is perfect for workers with dirty hands or wearing gloves
- Cord can be attached at any desired length
- Large soft foam pod for a comfortable, snug seal
Specifications
Yes | |
Universal | |
No | |
100 Pairs/Pk. | |
30 dB | |
Reusable |
Class AL | |
Construction, Utilities, Manufacturing, Production, Mining, Oil and Gas, and Other Various Industries | |
Poly bag | |
Foam | |
Bell | |
No |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title