Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ Hygiene Kit
SDP

Catalog No. 19120505 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Hygiene Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-120-505 Hygiene Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-120-505 Supplier Moldex™ Supplier No. 6105

Hygiene Kit For Moldex™ M1, M2, M3 and Z2 Ear Muffs include replacement ear cushions and foam inserts.

  • Cushions and foam inserts for the following Moldex Ear Muffs
    • M1 Premium
    • M2 Multi-Position
    • M3 Cap-Mounted Ear Muffs
    • Z2 Multi-Position Economy
  • 100% PVC-Free
  • Packaged 1 kit per pack and 20 packs per case

Specifications

Color Black
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Equipment) For hard hats
Headband Material Plastic
For Use With (Application) Hygiene Kits for M1, M2, M3, and Z2 Ear Muffs
Model 6105
Pairs per Package 1 Pk.
Noise Reduction Rating (NRR) 24 dB
Plug Shape Curvy
Product Type Hygiene Kit
Dielectric No
Includes Special molded grips
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.