Learn More
Moldex™ Mellows™ Disposable Ear Plugs
SDP
Mellows™ disposable ear plugs are made of extra-soft, extra-light foam and are appropriate for mid- to high-level noise areas.
Supplier:  Moldex™ 6820
 View more versions of this product
Catalog No. 19-384-341
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
  • Extra-soft, low-pressure foam for comfort and fit
  • Tapered shape for easy insertion and removal
  • Non-irritating smooth surface
  • High visibility color makes compliance checks quick and easy
  • Shaped to fit even very small ear canals
Specifications
Specifications
Yes
Universal
Yes
200 Pairs/Pk.
Orange
Uncorded
Single-use
Class AL
Construction, Utilities, Manufacturing, Production, Mining, Oil and Gas, and Other Various Industries
Poly bag
Foam
30 dB
Bullet
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.