Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ Z2 Multi-Position Economy Ear Muff
SDP

Catalog No. 191303463 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Z2, Multi-position
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-130-3463 Z2, Multi-position
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-130-3463 Supplier Moldex™ Supplier No. 6201

Z2 Ear Muffs feature a three-position adjustability with a lightweight plastic headband and special molded grips.

  • Can be worn over-the-head, behind-the-neck or under-the-chin
  • Molded grips make fitting and alignment easy and quick
  • Cup adjustment is highly variable and distributes pressure evenly for a comfortable fit
  • Lightweight plastic headband

Specifications

Color Black
ANSI S3.1974 Yes
Cushion Material Foam
Cushion Color Black
For Use With (Equipment) Moldex™ Hygiene Kit
Headband Material Plastic
For Use With (Application) Construction, Energy and Utilities, General Manufacturing, Mining, Oil and Gas, Transportation
Model 6201
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 25 dB
Product Type Z2 Series
Headband Color Black
Dielectric No
Material Plastic
Style Multi-position
Foldable No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.