Learn More
Moldex™ 1500 Series N95 Healthcare Particulate Respirators and Surgical Masks
SDP
Catalog No. 191500427 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Size:
Large
Low profile
Medium
Small
X-Small
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-150-0427 Small
19-150-050 X-Small
19-150-0428 Medium
19-150-0429 Large
19-150-0430 Low profile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-150-0427 Supplier Moldex™ Supplier No. 1511 N95
Add to Cart
Add to Cart
This item is not returnable. View return policy
N95 healthcare and surgical mask feature ASTM Level 3 protection (BFE >99.9%) and are ideal for use in healthcare settings and hospitals.
  • Molded nose bridge with soft foam nose cushion provides easy, comfortable, dependable fit
  • Dura-Mesh™ shell prevents filter media contact by contaminated gloves or hands; resists collapsing, even in heat and humidity
  • Nonlatex straps don't irritate or require prestretching
  • Highest ASTM™ level of fluid resistance on the market (160mmHg)
  • Bacterial filter efficiency >99.9%
  • Softspun™ inner lining is soft to the touch
  • Available in four color-coded sizes
  • ASTM Level 3
Order Info
20 respirators per pack.
Specifications
Aerosol Type N - Not Resistant to Oil
ANSI Standard ANSI/ISEA 110-2009, Section 7.11.1
Certifications/Compliance ASTM F1862
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Design Type Cup/Molded
Disposable Single-use
Filter Type Surgical N95
Material Dura-Mesh
Product Type Healthcare Particulate Respirator and Surgical Mask
Size Small
With Exhalation Valve No
Filter Efficiency 95%
Latex Free Yes
Material (Liner) Softspun™
Model 1511
Nose Piece Material Soft Foam
Nosepiece Type Molded Nose Bridge
Quantity 20/Box; 8 Boxes/Cs.
Resistant To Fluid 160 mmHg
Sterility Non-sterile
Strap Attachment Type Dual Elastic Straps
FDA Cleared Surgical Mask Yes