Description
- Wider field of vision because of low profile
- Encased in plastic for durability and rough handling
- Fits under welding helmets
- Easy twist on/twist off, bayonet style attachment
- Efficiency level of 99.97% or greater against both oil and non-oil based particulate aerosols
- Piggyback the 7940 filter disk with any Moldex™ gas/vapor cartridge using the 7920 Piggyback adaptor
- Meets heat and flame resistance in accordance with ANSI/ISEA 110-2003 Section 7.11.1
- 100% PVC-Free™
Specifications
Specifications
| Product Type | P100 Filter Disk |
| For Use With (Application) | Can be used for both oil and non-oil based particulates |
| Quantity | 1 Pair/Bag |