Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Moldex™ 8000 Series Pre-Assembled Multi-Gas Vapor Smart™ Cartridge Respirators
Pre-assembled respirator kits simplify inventory and are ideal for use against multiple hazards in the same workplace. Includes 8000 Series facepiece and 8600 Multi-Gas/Vapor Smart Cartridges.
$56.21 - $64.25
Specifications
Cartridge Type | Multi-Gas, Vapor Smart™ Cartridges |
---|---|
Certifications (NIOSH) | NIOSH 42 CFR Part 84, TC-23C-1223 |
Color | Gray |
Material | Thermoplastic Elastomer |
Product Type | Pre-Assembled Half-Mask Respirator |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-003-465A
|
Moldex™
8601 |
Small |
Each for $56.21
Case of 12 EA for $607.04
|
|
|||||
19-003-465B
|
Moldex™
8602 |
Medium |
Each for $64.25
Case of 12 EA for $692.00
|
|
|||||
19-003-465C
|
Moldex™
8603 |
Large | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
- 8000 Series half-mask pre-assembled with 8600 multi-gas/vapor smart cartridges
- Simplifies inventory by reducing need to stock multiple cartridge types
- Approved for nine common gas/vapors, replaces up to five gas/vapor cartridges
- Ideal for use against multiple hazards in same workplace
- For oil-free paint sprays and pesticides
- Soft, durable facepiece and lightweight design with extra-wide sealing
- Exhalation valve positioned for better communication
- Cartridges have a snap-in mount for easy replacement and no misthreading
- Living hinge allows head harness to be folded comfortably around neck when not in use
- Low profile, compact design fits well under welding helmets and with safety glasses
- Maintenance made simple with very few replacement parts
Specifications
Multi-Gas, Vapor Smart™ Cartridges | |
Gray | |
Pre-Assembled Half-Mask Respirator | |
8000 Facepiece, 8600 Multi-Gas, Vapor Smart™ Cartridges |
NIOSH 42 CFR Part 84, TC-23C-1223 | |
Thermoplastic Elastomer | |
Degreasing, forging, foundries, heat treating operations, paper making, rendering, rubber manufacturing |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title