Learn More
Moldex™ Probed cartridge kit for quantitative fit testing on 8000 series respirators.
SDP
Catalog No. 19013470 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Probed Cartridge Kit
Catalog No.
19-013-470 Probed Cartridge Kit
1 options
Catalog No. 19-013-470 Supplier Moldex™ Supplier No. 8006
Kit includes everything needed to perform quantitative fit testing using TSI Portacount on all 8000 Series Moldex™ respirators.
  • Comply with OSHA standards for quantitative fit testing
  • Includes:
    • 1 Probed Cartridge For Use With 8000 Series
    • 1 Pair 8940 P100 Filter Disks
    • 1 Pair 8920 Piggyback Adapters
    • 1 Pair 8900 Filter Disk Holders
    • Instructions and certificates
  • Respirators sold separately
Specifications
Product Type Probed Cartridge Kit
With Exhalation Valve No
For Use With (Application) Use to quantitatively fit test 8000 respirators with portacount™
For Use With (Equipment) 8000 respirators with portacount™
Model 8006
Quantity 1 Ea.