Learn More
Moldex™ Rock Band™ Canal Cap Hearing Protector
SDP
Catalog No. 191500135 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Ear band
Catalog No.
19-150-0135 Ear band
1 options
Catalog No. 19-150-0135 Supplier Moldex™ Supplier No. 6500
Two flexible arms distribute pressure evenly and the soft reusable pods gently insert into the ear canal. Exciting colors make the Rock Band™ Hearing Protector popular and more appealing with the new generation of workers.
  • Cooler and lighter alternative to ear muffs
  • Comfortable reusable pods gently cap off the ear canal
  • Available in 5 fun colors - blue, green, gray, red, yellow
  • Each box of 10 includes two of each color
  • Flexible band arms distribute pressure evenly for maximum comfort
  • Rock Band™ does not interfere with eyewear
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class BL
Size Universal
Headband Material Polycarbonate
For Use With (Application) For Intermittent Use When Switching Between Noisy and Quiet Areas
Pairs per Package 10 Bands/Pk.
Plug Material PVC-free
Plug Color Black
Noise Reduction Rating (NRR) 22 dB
Plug Shape Pod
Metal Detectable No
Headband Color Multicolor