Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ M1 Premium Folding Ear Muff
SDP

Catalog No. 191600154 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
M1 Series, Camouflage, Special Ops
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-160-0154 M1 Series, Camouflage, Special Ops
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-160-0154 Supplier Moldex™ Supplier No. 6109

The M1 Premium Ear Muff features a durable, black-chrome, spring steel headband that resists fatigue.

  • Six different adjustment points
  • Black-chrome, spring-steel headband resists fatigue and prevents it from twisting out of shape
  • Folds into a compact size for easy storage
  • SoftCoat™ covering helps muffle transmitted noise
  • Camouflage/Black

Specifications

Color Camouflage
ANSI S3.1974 Yes
Cushion Material Foam
Cushion Color Black
For Use With (Equipment) Moldex™ Hygiene Kit
Headband Material Chrome
For Use With (Application) Military, Armed Forces, Federal, Law Enforcement
Model 6109
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 29 dB
Product Type M1 Series
Headband Color Black
Dielectric No
Style Over-the-head
Foldable Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.