Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Disposable Screw Cap Closure

Catalog No. 02681129 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02-681-129 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-681-129

missing translation for 'validMainframeMsg'

Threaded cap ensures a secure seal and disposable for convenience

TRUSTED_SUSTAINABILITY

Specifications

Product Type Screw Cap
Color Blue
Quantity 2000/Cs.
For Use With (Equipment) 5 and 10 mL Transport Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.