Learn More
3M™ Molecular Detection Assay 2, Campylobacter Enrichment Broth
Shop All AquaPhoenix Scientific Products

Description
- Product Type: Microbiology Media
- Species Detected: Campylobacter
- Test Type: Food and Beverage, Environmental Test
- For Use With: 3M™ Molecular Detection System CE250 3M™ Campylobacter Enrichment Broth
- Application: Rapid and specific detection of Campylobacter in enriched food and environmental samples
- Quantity: 96 per pack
- Key Benefits:
- Pre-dispensed, ready-to-use reagents reduce prep time
- Fewer test steps and simple protocol minimize user error
- Color-coded assay tubes and matching software interface aid tracking and identification
- Simultaneous testing of up to 96 samples per run improves lab efficiency
- Real-time detection provides positive results in as little as 15 minutes
- Ideal for HACCP programs and food safety compliance
Specifications
Specifications
| Detects | Campylobacter |
| Product Type | Microbiology Media |
| Test Type | Food and Beverage, Environmental Test |
| For Use With (Application) | Detecting Campylobacter in enriched food and environmental samples |
| For Use With (Equipment) | CE250 3M™ Campylobacter Enrichment Broth, 3M™ Molecular Detection System |
| Quantity | 96/Pk. |
| Species | Campylobacter MDA2CAM96 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.