Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Molecular Detection Assay 2 - E. coli O157, Sold by AquaPhoenix Scientific
SCI_ED

Catalog No. S26893 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
96 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S26893 96 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S26893 Supplier 3M™ Supplier No. MDA2ECO96
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use with 3M™ Molecular Detection System for rapid and specific pathogen detection of E. coli O157, including H7, in enriched food samples

  • One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees
  • Ready-to-use and pre-dispensed reagents provide convenience and minimize chance of human error
  • Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification
  • Ability to run all assays simultaneously and flexibility to test up to 96 samples in each run helps operate more efficiently
  • Real-time amplification and detection process identifies positive results in as early as 15 min. and allows for critical decisions to be made quickly

Specifications

Detects E. coli O157 (including H7) MDA2ECO96
Product Type Microbiology Media
For Use With (Equipment) 3M™ Molecular Detection System
Quantity 96 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.