Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Molecular Detection Assay 2, Sold by AquaPhoenix Scientific
SCI_ED

Catalog No. S26896 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Detects:
Listeria monocytogenes species
Salmonella species
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detects
S26896 Salmonella species
S26895 Listeria monocytogenes species
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S26896 Supplier 3M™ Supplier No. MDA2SAL96
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used with 3M™ molecular detection system for rapid and specific detection of species in enriched food and environmental samples

  • One assay protocol and fewer steps than most tests reduces repetitive tasks
    • Easy to train employees on
  • Ready-to-use and pre-dispensed reagents provide convenience and minimize chance of human error
  • Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification
  • Ability to run all assays simultaneously and flexibility to test up to 96 samples in each run
    • Helps to operate more efficiently
  • Real-time amplification and detection process identifies positive results in as as little as 15 minutes
    • Able to make critical decisions faster

Specifications

Detects Salmonella species
Product Type Microbiology Media
Test Type Molecular Detection Assay
For Use With (Application) Food and environmental samples
For Use With (Equipment) 3M™ molecular detection system
Quantity 96 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.