Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel™ Molecular Influenza A+B Contol Set
SDP

Catalog No. 23044004 Shop All QuidelOrtho Products
Change view
Click to view available options
No. of Reactions:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
23-044-004 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-044-004 Supplier Quidel™ Supplier No. M106
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Influenza A+B molecular control set, 2 mL positive control, 2 mL negative control

  • Refrigeration storage
  • Ten reactions

Specifications

Format Kit
No. of Reactions 10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.