Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Molecular Orbital Theory Model Kit
SCI_ED

Catalog No. S139065 Shop All Walter Products Products
Click to view available options
Grade:
6+

Catalog No. S139065

Supplier: Walter Products™ MOLE1017

Only null left
Add to Cart
Add to Cart

Provide a visual representation of Molecular Orbital Theory with this model kit.

This colorful and durable set of molecular orbit models is a great tool for visualizing basic molecular orbital theory.

  • Molecular geometry and bond angles can be visibly seen in the structure, making it great for studying
  • Excellent molecular model kit for students studying chemistry in middle school or university
  • This set contains S, Px, Py, Pz, Sp, Sp2, and Sp3 molecular orbitals
  • Kit contains the standard S orbital and the 3 xyz P orbitals
  • Each model is mounted on a plastic base

Specifications

For Use With (Application) Chemistry
Grade 6+
Material Plastic
Product Type Model Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.