Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATTO-TAG™ CBQCA Derivatization Reagent (CBQCA; 3-(4-Carboxybenzoyl)quinoline-2-Carboxaldehyde)

Catalog No. A6222
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A6222 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A6222 Supplier Invitrogen™ Supplier No. A6222
Add to Cart
Edge
Add to Cart

Reacts specifically with primary amines to form conjugates that can be analyzed by electrophoretic or chromatographic methods

  • High sensitivity, freedom from background and long-wavelength excitability make these potential reagents for researcher, diagnostic and forensic applications
  • Resulting products of CBQCA are maximally excited at 450nm or by 442nm spectral line of He-Cd laser
  • Have emission maxima at approx. 550nm
  • In capillary zone electrophoresis (CZE), sensitivity of detection of ATTO-TAG CBQCA conjugates should be in attomole range (10-18 moles)

Anion Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Reactive Moiety Aldehyde
Shipping Condition Room Temperature
Form Solid
Product Line ATTO-TAG
Quantity 10 mg
For Use With (Application) Cell Viability, Proliferation and Function; Protein Electrophoresis and Western Blotting
Target Primary Amine
Type Derivatization Kits and Reagents
Label or Dye ATTO-TAG CBQCA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.