Learn More
Invitrogen™ ATTO-TAG™ FQ Amine-Derivatization Kit
Description
- High sensitivity, freedom from background and long-wavelength excitability make these potential reagents for researcher, diagnostic and forensic applications
- Resulting products of FQ are maximally excited at 480nm or by 488nm spectral line of argon-ion laser and have emission maxima at approximately 590nm
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Reactive Moiety | Aldehyde |
| Shipping Condition | Room Temperature |
| Product Line | ATTO-TAG |
| Quantity | 1 Kit |
| For Use With (Application) | Protein Electrophoresis and Western Blotting |
| Target | Primary Amine |
| Type | Derivatization Kits and Reagents |
| Label or Dye | ATTO-TAG FQ, ATTO-TAG FQ |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.