Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATTO-TAG™ FQ Derivatization Reagent (FQ; 3-(2-Furoyl)quinoline-2-Carboxaldehyde)

Catalog No. A10192
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10192 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10192 Supplier Invitrogen™ Supplier No. A10192
Add to Cart
Edge
Add to Cart

Allows ultrasensitive detection of primary amines

Recently, researchers described a simple method in which the ATTO-TAG FQ reagent was used to derivatize very dilute solutions (10-8 M) of peptides. Available as standalone product or as component of an amine-derivatization kit (A-2334). Particularly well-suited to derivatization of peptides, amino acids, aminated sugars and other amine-containing molecules for detection by capillary electrophoresis with laser-induced fluorescence, as its nonfluorescent in unreacted state and reacts specifically with amines to form highly fluorescent conjugates.

Anion Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Reactive Moiety Aldehyde
Shipping Condition Room Temperature
Form Solid
Product Line ATTO-TAG
Quantity 10 mg
For Use With (Application) Detection of primary amines
Target Primary Amine
Type Derivatization Kits and Reagents
Label or Dye ATTO-TAG FQ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.