Learn More
Invitrogen™ BacMam Enhancer Kit
Catalog No. B10107
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
B10107 1 kit
1 options
Catalog No. B10107 Supplier Invitrogen™ Supplier No. B10107
  • $196.00
    $160.72 / Each of 1
    Save $35.28
    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart
Includes
Each kit contains sufficient material for treating 10 X 96 microplates.
Helps to increase expression of BacMam reagents in mammalian cells
To use, simply add the BacMam enhancer to cell culture media with or without serum that have already been treated with the BacMam reagent, then incubate cells for 1.5–2 hours at 37°C and 5% CO2 and replace with fresh medium. Different cell types vary in their response to the BacMam enhancer and for best results the amount and treatment time will need to be optimized. Once prepared, the enhancer solution is stable to multiple freeze/thaw cycles following the recommended storage conditions. If desired, the solution can be divided into aliquots following reconstitution.
Cell Analysis, Cell Structure, Cell-Based Ion Channel Assays, Cytoplasm & Cytosol, Cytoskeleton, Drug Discovery & Development, Endoplasmic Reticulum (ER), Golgi, Ion Channel Biology, Lysosomes, Mitochondria, Mitochondrial Structure, Peroxisomes, Target & Lead Identification & Validation
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Each kit contains sufficient material for treating 10 X 96 microplates. Store at 2—6°C, do not freeze. When stored as directed the product is stable for 6 months.
Quantity 1 kit
Inhibitors Histone Deacetylase Inhibitor
Product Line Molecular Probes
Shipping Condition Room Temperature
Specific Inhibit or Target Histone deacetylase
Label Type Fluorescent Dye
Product Type BacMam Enhancer Kit
Sub Cellular Localization Actin, Endoplasmic Reticulum, Endosomes, Golgi, Mitochondria
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.