Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bis-BODIPY™ FL C11-PC) (1,2-Bis-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Undecanoyl)-sn-Glycero-3-Phosphocholine)

Catalog No. B7701
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B7701 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B7701 Supplier Invitrogen™ Supplier No. B7701
  • $409.00
    $335.38 / Each of 1

    Save $73.62

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Dye-labeled phospholipids used to monitor phospholipase A (PLA) activity

Molecular Probes phospholipase substrates are dye labeled phospholipids used to monitor phospholipase A (PLA) activity in purified enzyme preparations, cell lysates, and living cells. Two structural variations of these substrates are available, which confer specificity for PLA1 and PLA2. Fluorescently labeled versions of the cleavage product generated from these substrates are also available for use as standards for evaluating percentage substrate conversion in enzymatic reactions.

  • Label (Ex/Em): Bis-BODIPY FL C11-PC (approx. 488/530nm)
  • Specificity: PLA1 or PLA2
  • Cleavage product: D3862
  • Lyophilized product can be dissolved in ethanol for use
  • Fluorescence is measured either in microplate assays or using flow cytometry

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining and Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phospholipids
Quantity 100 μg
Physical Form Solid
Product Line BODIPY
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.