Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BODIPY™ 505/515 (4,4-Difluoro-1,3,5,7-Tetramethyl-4-Bora-3a,4a-Diaza-s-Indacene)

Catalog No. D3921
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3921 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3921 Supplier Invitrogen™ Supplier No. D3921
  • $323.00
    $264.86 / Each of 1

    Save $58.14

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

With its nonpolar structure and long-wavelength absorption and fluorescence, BODIPY™ 505/515 can be used as a stain for netural lipids and as a tracer for oil and other nonpolar lipids.

With its nonpolar structure and long-wavelength absorption and fluorescence, BODIPY™ 505/515 can be used as a stain for netural lipids and as a tracer for oil and other nonpolar lipids.

To make a concentrated stock solution from 1–5 mM, dissolve in either high quality anhydrous DMSO or 100% ethanol. Store frozen, desiccated and protected from light.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Shipping Condition Room Temperature
Product Line BODIPY
Label Type BODIPY Dyes
Molecular Formula C13H15BF2N2
Physical Form Solid
Product Type Fatty Acid
Excitation/Emission 502/510 nm
Molecular Weight (g/mol) 248.08

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.