Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BODIPY™ FL DHPE (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Propionyl)-1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt)

Catalog No. D3800
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3800 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3800 Supplier Invitrogen™ Supplier No. D3800
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Can form excimers at high concentrations

BODIPY DHPE has the green-fluorescent dye attached to the head group of the phospholipid. Unlike fluorophores like pyrene, DPH and NBD, BODIPY FL is relatively environment insensitive and is fluorescent in both aqueous and lipid environments.

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phospholipids
Quantity 100 μg
Physical Form Solid
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.