Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BODIPY™ TR Ceramide

Catalog No. D7540
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D7540 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D7540 Supplier Invitrogen™ Supplier No. D7540
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Red-fluorescent dye

BODIPY TR ceramide can be used to stain Golgi apparatus in live cells.

Cell Analysis, Cell Structure, Golgi

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sphingolipids
Quantity 250 μg
Physical Form Solid
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.