Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BODIPY™ TR Ceramide complexed to BSA

Catalog No. B34400
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34400 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34400 Supplier Invitrogen™ Supplier No. B34400
  • $457.00
    $374.74 / Each of 1

    Save $82.26

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Can be used in sphingolipid transport and metabolism mechanisms

The red-fluorescent BODIPY TR C5 ceramide complexed to bovine serum albumin. Complexing fluorescent lipids with BSA facilitates cell labeling by eliminating the need for organic solvents to dissolve the lipophilic probe; the BSA-complexed probe can be directly dissolved in water.

Cell Analysis, Cell Structure, Golgi

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sphingolipids
Quantity 5 mg
Physical Form Solid
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.