Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cholera Toxin Subunit B (Recombinant), Alexa Fluor™ 555 Conjugate

Catalog No. C22843
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22843 500 μg
C34776 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22843 Supplier Invitrogen™ Supplier No. C22843
Add to Cart
Edge
Add to Cart

Made from a recombinant version of the B subunit only to provide a very high-purity product that is completely free of the toxic A subunit

  • Cholera toxin B subunit (CT-B) attaches to cells by binding to ganglioside GM1, making it powerful tool for retrograde labeling of neurons
  • This tracer has been used in variety of applications, including tracing of rat forebrain afferents, projections of the parabrachial region, and neurons of the urinary bladder wall
  • When used in neuronal tracing applications, CT-B is typically introduced by pressure injection or by iontophoretic injection into neural tissue
  • At neutral pH, 11.4 kDa B subunit exists as 57 kDa pentamer
  • Lyophilized product can be dissolved in buffer (e.g., PBS) for use
  • More recently, researchers have found that CT-B can be used as marker for lipid rafts, which are membrane microdomains enriched in cholesterol and sphingolipids thought to be important in cell signaling
  • For lipid raft staining, cells are first incubated with fluorescent CT-B, then an anti-CT-B antibody is added to crosslink the CT-B in the lipid rafts into distinct patches on the plasma membrane
  • These patches are easily visualized by fluorescence microscopy

  • Label (Ex/Em): Alexa Fluor™ 555 (555/565nm)
  • At neutral pH, the 11.4 kDa B subunit exists as a 57 kDa pentamer

Cell Analysis, Cell Structure, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Lipid Rafts, Microbial Tracking, Neuronal Tracing

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 μg
Conjugate Alexa Fluor 555
Recombinant Recombinant
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Protein Form Recombinant
Form Lyophilized
Protein Subtype Cholera Toxin
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.