Learn More
Invitrogen™ Deoxyribonuclease I, Alexa Fluor™ 488 Conjugate
Description
The bright green fluorescent Alexa Fluor 488 DNase I selectively binds to G-actin (globular).
Cell Analysis, Cell Structure, Cytoskeleton
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Quantity | 5 mg |
| Conjugate | Alexa Fluor 488 |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Excitation/Emission | 495⁄519 |
| Label or Dye | Alexa Fluor™ 488 |
| Label Type | Alexa Fluor Dyes |
| Shipping Condition | Room Temperature |
| Sub Cellular Localization | Actin, Cytoskeleton |
| Form | Lyophilized |
| Product Line | Alexa Fluor |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.