Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DiD' oil; DiIC18(5) oil (1,1'-Dioctadecyl-3,3,3',3'-Tetramethylindodicarbocyanine Perchlorate)

Catalog No. D307
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D307 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D307 Supplier Invitrogen™ Supplier No. D307
  • $312.00
    $255.84 / Each of 1

    Save $56.16

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Longer-wavelength DiI analog

The far-red fluorescent, lipophilic carbocyanine DiD is an oil at room temperature and weakly fluorescent in water but highly fluorescent and quite photostable when incorporated into membranes. It has an extremely high extinction coefficient and short excited-state lifetimes (∽1 nanosecond) in lipid environments. Once applied to cells, the dye diffuses laterally within the plasma membrane.

Cell Analysis, Cell Tracing & Tracking, Neuronal Tracing

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Content And Storage Store at room temperature and protect from light.
Excitation Wavelength Range 644 nm
For Use With (Equipment) Fluorescence Microscope
Quantity 25 mg
Detection Method Fluorescence
Emission 665 nm
Shipping Condition Room Temperature
Product Type Liphophilic Tracer
Sub Cellular Localization Cell Membranes, Lipids
Molecular Weight (g/mol) 959.92
Show More Show Less
I'm labeling live cells with Vybrant DiI or DiD lipophilic cyanine dyes. DiI gives a nice even membrane labeling, but DiD is more "spotty". What can be done?

This is expected. DiD (which is far-red fluorescent) is never as uniform as DiI (which is orange fluorescent). If uniformity is desired, try increasing the label time and concentration, but it still isn't likely to be as uniform as DiI. CellMask Deep Red Plasma Membrane stain is much more uniform and is about the same wavelength as DiD. However, if you intend to do cell tracking over days, CellMask stain has not been tried for that application.

I stained my cells with a lipophilic cyanine dye, like DiI, but the signal was lost when I tried to follow up with antibody labeling. Why?

Since these dyes insert into lipid membranes, any disruption of the membranes leads to loss of the dye. This includes permeabilization with detergents like Triton X-100 or organic solvents like methanol. Permeabilization is necessary for intracellular antibody labeling, leading to loss of the dye. Instead, a reactive dye such as CFDA SE should be used to allow for covalent attachment to cellular components, thus providing for better retention upon fixation and permeabilization.

How long does it take for lipophlic tracers to transport along the membrane? How much faster are the FAST lipophilic dyes?

The transport is fairly slow, around 6 mm/day in live tissue and slower in fixed tissue, so diffusion of lipophilic carbocyanine tracers from the point of their application to the terminus of a neuron can take several days to weeks The FAST DiO and DiI analogs (which have unsaturated alkyl tails) can improve transport rate by around 50%.

Which form of the lipophilic tracers (DiO, DiI, DiD, etc) should I use?

Select the dye that is compatible with your available excitation source(s) and emission filter set/channels. The solid, paste and crystal forms can be applied directly to neurons in tissues. For labeling cells in culture or microinjection, the lipophilic dyes in solution or solid form can be used.

I want to label two cell populations and then perform a cell fusion assay. Which reagents are best for imaging this?

Lipophilic cyanine dyes are preferred for this sort of assay, since they insert into cellular membranes and then, upon fusion, are shared by the fused cells as the membranes are shared. For example, one cell population can be labeled with DiI (orange-red) and another cell population can be labeled with DiO (green), and when the cells fuse, the combined color appears yellow (when imaged with a dual-bandpass filter set).

I want to track my cells with a nucleic acid stain, like DAPI or Hoechst dye. Do you recommend this?

This is not recommended. When these stains bind to DNA and RNA, they may affect the normal function of the nucleic acids, disrupting transcription, as well as replication. Other reagents, such as CellTracker dyes or Qtracker reagents are more optimized for tracking without disrupting normal activity. If a nuclear label is still desired, though, and the cells are mammalian and non-hematopoietic, CellLight nuclear reagents can transiently transfect cells to express GFP or RFP on a nuclear-expressing protein for up to several days without affecting function.

I want to track my cells over time, and you have a lot of options to choose from. How do I pick the right one?

Please see this Web link (http://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/cell-tracking.html) to help you choose the right option for your application. Start by planning how long you want to track your cells, then consider the mechanism of binding. Calcein dyes are very uniform in label and are good for short-term cell migration, but may be rapidly effluxed from some cell types. Lipophilic cyanine dyes, such as DiI, DiO, and similar dyes label cell membranes, don’t disrupt function, and can last longer, but have the potential to cross to other cells if membranes fuse. They are also lost upon permeabilization. CellTracker dyes are better for longer-term labeling, as they possess a mildly reactive chloromethyl moiety that allows covalent binding to cellular components. CFDA SE also covalently binds to cellular components. With all the reagents, their retention within cells is dependent upon the rate of cell division and the inherent properties of the cell (active efflux, membrane and protein turnover rates, etc.) and reagents that allow for covalent attachment exhibit longer retention than those that do not.

The longest-lasting and brightest options are the Qtracker reagents, which are taken up through endocytosis. These are so bright individual quantum dots can be detected, and are also robust enough to survive not only fixation and permeabilization, but even the heat and solvents used in paraffin processing.


WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.