Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DQ™ Collagen, type IV From Human Placenta, Fluorescein Conjugate

Catalog No. D12052
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12052 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12052 Supplier Invitrogen™ Supplier No. D12052
Add to Cart
Edge
Add to Cart

Analog of the natural substrate

The fluorogenic DQ™ collagen can be used to directly monitor collagenase activity. DQ™ substrates are analogs of the natural substrate that have an excessive number of fluorescent dyes attached so that the fluorescence signal is almost non-existent. This quenching of the signal is caused by the close proximity of the dyes on the intact substrate. The enzyme-driven hydrolysis of the substrate results in separation of the dye results in separation of the dye molecules from one another and the fluorescence signal increases.

Cell Adhesion, Cell Analysis, Cell Viability, Proliferation and Function, Chemotaxis and Cell Migration, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Form Lyophilized
Product Line DQ
Quantity 1 mg
Substrate Protease Substrate
Substrate Properties Protein-Based Substrate
Target Enzyme Metalloproteinase
Shipping Condition Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.