Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FilmTracer™ FM™ 1-43 Green Biofilm Cell Stain

Catalog No. F10317
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10317 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10317 Supplier Invitrogen™ Supplier No. F10317
  • $546.00
    $447.72 / Each of 1

    Save $98.28

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Lipophilic styryl compounds suitable for wide variety of studies involving plasma membrane and vesiculation

The water-soluble FM dyes are virtually nonfluorescent in aqueous media and are believed to insert into the surface membrane where they become intensely fluorescent. FM 1-43 stain has been used successfully to stain the cell bodies specifically in a complex biofilm mileu, including Pseudomonas aeruginosa, Escherichia coli, Staphylococcus sp., Acidothiobacillus caldus, and Vibrio cholerae.

  • Flow Cytometer Laser Lines: 488
  • Platform: Confocal Microscope, Flow Cytometer, Fluorescence Microscope, Fluorescent Imager, Microplate Reader
  • Label or Dye: FM 1-43/lipid

Analysis of Bacteria, Bacterial Staining & Detection, Bacterial Tracing/Tracking/Morphology, Cell Analysis, Microbiological Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Content And Storage Store at room temperature; Dessicate; Protect from light
Excitation Wavelength Range 472⁄580
Dye Type FM 1-43⁄lipid
For Use With (Equipment) Confocal Microscope, Fluorescence Microscope, Flow Cytometer, Microplate Reader, Fluorescent Imager
Quantity 1 mg
Detection Method Fluorescence
Emission Visible
Form Solid
Product Line FilmTracer, Molecular Probes
Shipping Condition Room Temperature
Label Type Other Labels or Dyes
Product Type Dye
Sub Cellular Localization Cell Membranes, Lipids
Show More Show Less
The product sheet for FM 1-43 Dye states that the maximum for excitation is 510 nm and emissions is 626 nm, but the spectrum on the product page shows a peak at 470 nm and 580 nm. Can you clarify what the excitation and emission maxima is for this dye?

FM lipophilic styryl dyes are known to shift their spectral properties upon binding to membranes. These dyes exhibit an appreciable solvent and environmental dependency in both free and membrane-bound forms. The emissions of the membrane-bound dye shifts to a shorter wavelength compared to the emission in the solvent.

In solvent, FM 1-43 Dye (Cat. Nos. T35356, T3163, F35355, F10317) absorbs in the range of fluorescein (~490 nm) and initially emits at around 626-636 nm. However, upon membrane integration, it exhibits emission in the 565-590 nm range. Because this dye exhibits a large Stokes shift, it does not perfectly fit into a green or orange fluorophore profile, but is rather a yellow-fluorescent dye, and may require a long-pass emission filter.

Note: FM 1-43 Dye is often used with typical fluorescein or GFP optical filter sets for biological imaging but is poorly excited through tetramethylrhodamine filters.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.