Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FluoroMyelin™ Red Fluorescent Myelin Stain - Solution in Water

Catalog No. F34652
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F34652 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F34652 Supplier Invitrogen™ Supplier No. F34652
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

For use with fluorescence microscopy

The FluoroMyelin Red fluorescent myelin stain enables quick and selective labeling of myelin in brain cryosections in a single 20-minute labeling step plus washes. This stain can be used in conjunction with antibodies and other dyes, and with standard histochemical methods for cryosection material. Solution in water.

Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Neuronal Tracing

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Detection Method Fluorescence
Product Type Red Fluorescent Myelin Stain
Quantity 1 mL
Content And Storage • Provided as 300X concentration in water.
• When stored at 4°C, stable for at least 6 months after receipt.
• Protect from light.
Label or Dye FluoroMyelin™ Red
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope
Sub Cellular Localization Myelin
Excitation/Emission 558/654 nm
Product Line FluoroMyelin
Show More Show Less
I used one of your FluoroMyelin stains and noticed it stains cells other than glial cells. Is there something wrong with the product?

FluoroMyelin is a lipid stain, any lipid can be stained by it but there is a higher lipid content in myelin that it will stain much more intensely than other membranes.

Can the FluoroMyelin stains be used on neurons in culture?

We have only tested them on mouse brain cryosections. They might work on neurons in culture, but would likely stain the membrane and there would be some endocytosis of the stains. Myelin sheaths may show enhanced intensity.

Can the FluoroMyelin myelin stains be used on paraffin sections?

We have only tested them on mouse brain cryosections.

What are the FluoroMyelin myelin stains?

They are proprietary lipid stains that stain myelin, but are not neuron specific.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.