Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Muscimol, BODIPY™ TMR-X Conjugate

Catalog No. M23400
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23400 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M23400 Supplier Invitrogen™ Supplier No. M23400
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Muscimol is a powerful agonist of the GABAA receptor and has been widely used to reversibly inactivate localized groups of neurons.

Muscimol is a powerful agonist of the GABAA receptor and has been widely used to reversibly inactivate localized groups of neurons. The BODIPY TMR-X muscimol conjugate, can be used, in place of radioactive reagents and methods, to map the distribution of muscimol in the nervous system, as well as to detect the presence of GABAA receptors on cell surfaces.

To make a concentrated stock solution, dissolve in either H2O, buffer or normal saline (∼1 mM) or high quality anhydrous DMSO (∼2 mM). DMSO stock solutions should be stored frozen, desiccated and protected from light. Aqueous stock solution should be used fresh.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope
Detection Method Fluorescence
Product Line BODIPY
Molecular Formula C31H36BF2N5O5
Physical Form Solid
Color Yellow
Product Type Agonist
Excitation/Emission 543/572 nm
Molecular Weight (g/mol) 607.46
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.