Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NucBlue™ Live ReadyProbes™ Reagent (Hoechst 33342)

Catalog No. R37605
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R37605 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R37605 Supplier Invitrogen™ Supplier No. R37605
  • $205.00
    $188.65 / Each of 1

    Save $16.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Hoechst stain formulated in room temperature-stable solution that is provided in dropper bottle

Hoechst 33342 is perhaps the most ubiquitous nuclear fluorescence stain for live and fixed cells and tissue sections. The cell permeable stain binds preferentially to AT regions of DNA. It is excited by UV light and emits blue fluorescence at 460nm when bound to DNA.

  • Ready-to-use liquid formulation of high-purity Hoechst 33342
  • Typically Hoechst 33342 is supplied as solid or highly concentrated solution that requires weighing out and diluting several thousand-fold before use, and recommended storage is generally in freezer or refrigerator
  • This version is ready to use and can be stored next to a microscope or workstation. A liquid formulation in ultra-convenient dropper bottle allows you to see
  • Spectral properties of Hoechst 33342 (2'-[4-ethoxyphenyl]-5-[4-methyl-1-piperazinyl]-2,5'-bi-1H-benzimidazole), including a large Stokes shift, make it ideal for use with green (Alexa Fluor 488, FITC, GFP) and red (Alexa Fluor 594, Texas Red, rhodamine, mCherry, mKate-2) fluorophores in multicolor experiments
  • Because of its high affinity to DNA, Hoechst 33342 is also frequently used in cell counting, cell cycle, and cell replication studies to distinguish condensed nuclei in apoptotic cells, for cell-cycle studies, as a nuclear segmentation tool in high content imaging analysis, and to sort cells based on their DNA content
  • May be added directly to cells in full media, or buffer solutions
  • In most cases 2 drops/mL and incubation of 15 to 30 minutes will give bright nuclear staining; however, optimization may be needed for some cell types, conditions, and applications
  • In most cases, staining intensity increases with time if cells are not washed prior to imaging.
  • Excited by UV light at 360nm when bound to DNA, with an emission maximum at 460nm
  • Detected through blue/cyan filter

Apoptosis, Cell Analysis, Cell Cycle, Cell Proliferation, Cell Structure, Cell Tracing and Tracking, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, DNA Fragmentation, Flow Cytometry, Flow Cytometry Staining by Cell Structure, Immunofluorescence (IF), Immunofluorescence Counterstaining, Mounting and Fade Prevention, Immunofluorescence Staining and Detection, Nucleus, Nucleoli and Nuclear Envelope, Organelle Tracing

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Content And Storage 6 × 2.5 mL dropper bottles

Store at ≤ 25°C

Excitation Wavelength Range 360⁄460
Dye Type Hoechst 33342
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Quantity 1 kit
Detection Method Fluorescence
Emission Visible
Form Liquid
Product Line Molecular Probes
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Nucleic Acid Stain
Sub Cellular Localization Nucleus
Show More Show Less
Can I use the ReadyProbes reagents for flow cytometry?

This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.

What is the difference between NucBlue Live reagents stain and NucGreen Dead reagents stain?

The NucBlue Live reagent stains the nuclei of all cells, while NucGreen Dead reagent stains only the nuclei of dead cells.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.