Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ O-Rings for Attofluor™ Cell Chamber

Catalog No. O14804
Encompass_Preferred
Change view
Click to view available options
Quantity:
10⁄pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O14804 10⁄pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O14804 Supplier Invitrogen™ Supplier No. O14804
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Replacement part for durable and practical coverslip holder for viewing live-cell specimens on upright or inverted microscopes

Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunohistochemistry (IHC)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Type O-ring
Product Line Attofluor
Content And Storage Store at room temperature.
Quantity 10⁄pack
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.